ADV390216538 AChemBlock

1H-1,2,4-Triazole-3-sulfonamide

  • MDL number MFCD18207305
  • Cas Number : 89517-96-4
  • Molecular Weight : 148.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-1-2-4-triazole-3-sulfonamide.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3615
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart