ADV390216554 AChemBlock

3-Bromo-4-nitropyridine-N-oxide

  • MDL number MFCD00085925
  • Cas Number : 1678-49-5
  • Molecular Weight : 219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-nitropyridine-n-oxide.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P272;P280;P301+312;P302+352;P304+340;P305+351+338;P330;P333+313;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3703
pictograms GHS07
hstatements H302;H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart