ADV390216572 AChemBlock

6-Bromo-2-chloro-8-methoxy-quinazoline

  • MDL number MFCD09907930
  • Cas Number : 953039-14-0
  • Molecular Weight : 273.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-chloro-8-methoxy-quinazoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2813
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart