ADV390216573 AChemBlock

6-Bromo-2,4-dichloroquinazoline

  • MDL number MFCD09744007
  • Cas Number : 102393-82-8
  • Molecular Weight : 277.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-4-dichloroquinazoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2817
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart