ADV390216588 AChemBlock

2-Amino-5-bromo-benzenethiol

  • MDL number MFCD00839467
  • Cas Number : 23451-95-8
  • Molecular Weight : 204.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-bromo-benzenethiol.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-873
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart