ADV390216597 AChemBlock

2,4-Dibromo-[1,8]naphthyridine

  • MDL number MFCD14705080
  • Cas Number : 54569-27-6
  • Molecular Weight : 287.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dibromo-1-8-naphthyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-960
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart