ADV390216601 AChemBlock

4-Iodo-1-methylimidazole

  • MDL number MFCD01632214
  • Cas Number : 71759-87-0
  • Molecular Weight : 208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-iodo-1-methylimidazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-967
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart