ADV390216625 AChemBlock

3-Fluoro-2-nitroaniline

  • MDL number MFCD07368750
  • Cas Number : 567-63-5
  • Molecular Weight : 156.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-2-nitroaniline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=E-3042
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart