ADV390216649 AChemBlock

Pyrimidine-5-carboxaldehyde

  • MDL number MFCD03426065
  • Cas Number : 10070-92-5
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/pyrimidine-5-carboxaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-903
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart