ADV390216679 AChemBlock

tert-Butyl 1,9-Diazaspiro[5.5]undecane-9-carboxylate

  • MDL number MFCD09998745
  • Cas Number : 1031927-14-6
  • Molecular Weight : 254.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-9-diazaspiro-5-5-undecane-9-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4168
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart