ADV390216682 AChemBlock

1,8-Diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester

  • MDL number MFCD11223538
  • Cas Number : 1086394-59-3
  • Molecular Weight : 254.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-8-diazaspiro-5-5-undecane-8-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4171
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart