ADV390216684 AChemBlock

1-Oxa-8-azaspiro[4.5]decane hydrochloride

  • MDL number MFCD11042294
  • Cas Number : 3970-79-4
  • Molecular Weight : 177.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-oxa-8-azaspiro-4-5-decane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4176
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart