ADV390216689 AChemBlock

2-(Oxetan-3-yl)acetic acid

  • MDL number MFCD19442256
  • Cas Number : 1310381-54-4
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-oxetan-3-yl-acetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4181
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart