ADV390216690 AChemBlock

trans-tert-Butyl 3-Iodocyclobutylcarbamate

  • MDL number MFCD22415232
  • Cas Number : 1389264-19-0
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-3-iodocyclobutylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4182
pictograms GHS07; GHS09
hstatements H302;H410
Availability details for
Quantity
  • Availability
  • Add to Cart