ADV390216698 AChemBlock

4,6-Dichloro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD09880157
  • Cas Number : 5912-18-5
  • Molecular Weight : 187.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4191
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart