ADV390216704 AChemBlock

(R)-tert-Butyl 2-Cyanopiperazine-1-carboxylate

  • MDL number MFCD21364722
  • Cas Number : 1359658-44-8
  • Molecular Weight : 211.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-tert-butyl-2-cyanopiperazine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4197
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart