ADV390216721 AChemBlock

4-(Boc-amino)tetrahydropyran-4-carboxylic Acid

  • MDL number MFCD02683136
  • Cas Number : 172843-97-9
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-boc-amino-tetrahydropyran-4-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4215
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart