ADV390216724 AChemBlock

(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-5-yl)methanol

  • MDL number MFCD20687490
  • Cas Number : 1207543-24-5
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7h-pyrrolo-2-3-d-pyrimidin-5-yl-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4218
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart