ADV390216726 AChemBlock

2-Cyclopropyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD08686635
  • Cas Number : 886779-93-7
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-cyclopropyl-piperazine-1-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4219
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart