ADV390216731 AChemBlock

tert-Butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD08691407
  • Cas Number : 114214-49-2
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-oxa-3-azabicyclo-3-1-0-hexane-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4225
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart