ADV390216736 AChemBlock

2,6-Dichloro-5-fluoropyridine-3-carboxamide

  • MDL number MFCD01571365
  • Cas Number : 113237-20-0
  • Molecular Weight : 209.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloro-5-fluoropyridine-3-carboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4230
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart