ADV390216752 AChemBlock

N-Boc-(2)-(bromomethyl)morpholine

  • MDL number MFCD20489326
  • Cas Number : 765914-78-1
  • Molecular Weight : 280.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n-boc-2-bromomethyl-morpholine.html

Safety Information

signalword Danger
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4246
pictograms GHS05; GHS06; GHS07
hstatements H301;H312;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart