ADV390216766 AChemBlock

6-(Ethyloxycarbonyl)spiro[3.3]heptane-2-carboxylic acid

  • MDL number MFCD22419340
  • Cas Number : 28114-90-1
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-ethyloxycarbonyl-spiro-3-3-heptane-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4260
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart