ADV390216770 AChemBlock

2-(Diphenylmethyl)-2,6-diazaspiro[3.4]octane

  • MDL number MFCD16987815
  • Cas Number : 1250443-61-8
  • Molecular Weight : 278.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-diphenylmethyl-2-6-diazaspiro-3-4-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4264
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart