ADV390216775 AChemBlock

1H-Pyrazolo[4,3-b]pyridin-5-amine

  • MDL number MFCD22550615
  • Cas Number : 1206974-46-0
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-4-3-b-pyridin-5-amine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4269
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart