ADV390216788 AChemBlock

N-(2-(3-(1-Piperazinylmethyl)imidazo(2,1-b)thiazol-6-yl)phenyl)-2-quinoxalinecarboxamide

  • MDL number MFCD14584468
  • Cas Number : 925434-55-5
  • Molecular Weight : 469.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96.00
Storage temp. Store at Room Tem.
Physical Form off-white to brown solid
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/n-2-3-1-piperazinylmethyl-imidazo-2-1-b-thiazol-6-yl-phenyl-2-quinoxalinecarboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4282
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart