ADV390216789 AChemBlock

5-Bromo-3-methyl-7-azaindole

  • MDL number MFCD15529125
  • Cas Number : 1111637-94-5
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-methyl-7-azaindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4283
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart