ADV390216790 AChemBlock

(S)-tert-Butyl 3-methyl-1,4-diazepane-1-carboxylate

  • MDL number MFCD09840678
  • Cas Number : 194032-32-1
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form light-yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-tert-butyl-3-methyl-1-4-diazepane-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4284
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart