ADV390216794 AChemBlock

2-Amino-4-bromopyrimidine

  • MDL number MFCD09999168
  • Cas Number : 343926-69-2
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-4-bromopyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2371
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart