ADV390216817 AChemBlock

5-Chloro-3-cyclopropylpyrazole

  • MDL number MFCD17214378
  • Cas Number : 1416437-25-6
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-cyclopropylpyrazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5010
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart