ADV390216820 AChemBlock

(S)-1-Tetrahydrofuran-3-ylmethanamine hydrochloride

  • MDL number MFCD20526350
  • Cas Number : 1403763-27-8
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-1-tetrahydrofuran-3-ylmethanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5015
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart