ADV390216823 AChemBlock

[1,2,4]Triazolo[4,3-a]pyridine-8-carboxylic acid

  • MDL number MFCD07377043
  • Cas Number : 1216218-95-9
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-4-triazolo-4-3-a-pyridine-8-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5017
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart