ADV390216826 AChemBlock

7-Aminomethyl-2-methylindazole hydrochloride

  • MDL number MFCD22421660
  • Cas Number : 1400764-43-3
  • Molecular Weight : 197.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-aminomethyl-2-methylindazole-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5022
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart