ADV390216832 AChemBlock

(3aR,7aR)-2-Methyl-octahydro-3H-pyrrolo[3,4-c]pyridin-3-one

  • MDL number MFCD17016078
  • Cas Number : 1400744-16-2
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3ar-7ar-2-methyl-octahydro-3h-pyrrolo-3-4-c-pyridin-3-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5030
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart