ADV390216839 AChemBlock

5-Aza-spiro[3.5]nonane hydrochloride

  • MDL number MFCD23106462
  • Cas Number : 1419101-47-5
  • Molecular Weight : 161.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-aza-spiro-3-5-nonane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5037
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart