ADV390216845 AChemBlock

3-Fluoro-4-nitropyridine-N-oxide

  • MDL number MFCD00234027
  • Cas Number : 769-54-0
  • Molecular Weight : 158.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow crystal
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-4-nitropyridine-n-oxide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2430
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart