ADV390216854 AChemBlock

2-Cyano-3-cyclopropyl-acrylic acid

  • MDL number MFCD22377851
  • Cas Number : 98895-60-4
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-cyano-3-cyclopropyl-acrylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4313
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart