ADV390216859 AChemBlock

1-Boc-6-hydroxy-1-azaspiro[3.3]heptane

  • MDL number MFCD22415220
  • Cas Number : 1363381-56-9
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-6-hydroxy-1-azaspiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5053
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart