ADV390216868 AChemBlock

2-Boc-2-azaspiro[3.5]nonan-7-one

  • MDL number MFCD22566155
  • Cas Number : 1363381-22-9
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-boc-2-azaspiro-3-5-nonan-7-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5068
pictograms GHS07
hstatements H302;H313;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart