ADV390216869 AChemBlock

1-Cbz-1,8-diazaspiro[4.5]decane hydrochloride

  • MDL number MFCD20809860
  • Cas Number : 1385696-68-3
  • Molecular Weight : 310.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-1-8-diazaspiro-4-5-decane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5069
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart