ADV390216880 AChemBlock

trans-3-Methylsulfonylcyclobutylamine hydrochloride

  • MDL number MFCD22628711
  • Cas Number : 1408075-97-7
  • Molecular Weight : 185.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-3-methylsulfonylcyclobutylamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5076
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart