ADV390216893 AChemBlock

Methyl 5-bromo-4-nitro-1H-indazole-3-carboxylate

  • MDL number MFCD23106303
  • Cas Number : 1403767-11-2
  • Molecular Weight : 300.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-4-nitro-1h-indazole-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5089
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart