ADV390216912 AChemBlock

2-[1-(Cbz-amino)ethyl]-5-thiazolecarboxylic acid

  • MDL number MFCD20126213
  • Cas Number : 1095823-52-1
  • Molecular Weight : 306.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-1-cbz-amino-ethyl-5-thiazolecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5114
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart