ADV390216924 AChemBlock

6,7-Dimethoxy-1H-quinazolin-4-one

  • MDL number MFCD01570147
  • Cas Number : 13794-72-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white to yellow-brown powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-7-dimethoxy-1h-quinazolin-4-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5131
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart