ADV390216933 AChemBlock

3-[4-(Trifluoromethyl)phenyl]-pyrrolidine hydrochloride

  • MDL number MFCD03701254
  • Cas Number : 1095545-12-2
  • Molecular Weight : 251.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-trifluoromethyl-phenyl-pyrrolidine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5149
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart