ADV390216957 AChemBlock

Methyl 7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate

  • MDL number MFCD17015873
  • Cas Number : 1095822-17-5
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-7h-pyrrolo-2-3-d-pyrimidine-4-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5177
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart