ADV390216959 AChemBlock

4-Chloro-6-methyl-thieno[3,2-d]pyrimidine

  • MDL number MFCD17015877
  • Cas Number : 108134-22-1
  • Molecular Weight : 184.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-methyl-thieno-3-2-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5184
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart