ADV390216963 AChemBlock

1,2,3,5-Tetra-O-benzoyl-2-C-methyl-b-D-ribofuranose

  • MDL number MFCD07369658
  • Cas Number : 15397-15-6
  • Molecular Weight : 580.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-5-tetra-o-benzoyl-2-c-methyl-b-d-ribofuranose.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5196
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart