ADV390216964 AChemBlock

Methyl cis-3-amino-cyclobutanecarboxylate hydrochloride

  • MDL number MFCD11099895
  • Cas Number : 1212304-86-3
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-cis-3-amino-cyclobutanecarboxylate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5199
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart