ADV390216971 AChemBlock

3-Iodo-4,7-diazaindole

  • MDL number MFCD19382469
  • Cas Number : 889451-26-7
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-4-7-diazaindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5210
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart